Assay Details
| Gene Symbol: Fscn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172802.4 | NP_766390.2 | 1 - 2 | 1001 | 60 | |
| GenBank mRNA | AK044556.1 | - | 2 - 3 | 932 | 60 | |
| AK050002.1 | - | 1 - 2 | 1002 | 60 | ||
| BC109356.1 | - | 1 - 2 | 962 | 40057445 | 60 | |
| BC109357.1 | - | 1 - 2 | 962 | 40057447 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fscn2
Genomic Map