Assay Details
| Gene Symbol: Mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010813.3 | NP_034943.3 | 4 - 5 | 1215 | 100 | |
| GenBank mRNA | AK139743.1 | - | 3 - 4 | 237 | 100 | |
| AK147933.1 | - | 4 - 5 | 1199 | 100 | ||
| BC054534.1 | - | 4 - 5 | 1141 | 5721249 | 100 | |
| U77356.1 | - | 4 - 5 | 1215 | 100 | ||
| Y07609.1 | - | 4 - 5 | 1161 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mnt
Genomic Map