Assay Details
| Gene Symbol: Mettl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026197.3 | NP_080473.1 | 4 - 5 | 594 | 117 | |
| GenBank mRNA | AK011780.1 | - | 4 - 5 | 595 | 117 | |
| AK012739.1 | - | 4 - 5 | 595 | 117 | ||
| AK013306.1 | - | 4 - 5 | 606 | 117 | ||
| AK029045.1 | - | 4 - 5 | 578 | 117 | ||
| AK145022.1 | - | 4 - 5 | 602 | 117 | ||
| AK145662.1 | - | 4 - 5 | 594 | 117 | ||
| AK168798.1 | - | 4 - 5 | 544 | 117 | ||
| BC003467.1 | - | 4 - 5 | 565 | 117 | ||
| BC022902.1 | - | 4 - 5 | 606 | 117 | ||
| BC037452.1 | - | 4 - 5 | 543 | 117 | ||
| BC049897.1 | - | 4 - 5 | 523 | 3979938 | 117 | |
| BC054724.1 | - | 4 - 5 | 530 | 5695738 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl16
Genomic Map