Assay Details
| Gene Symbol: Cluh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081158.2 | NP_001074627.1 | 10 - 11 | 2056 | 60 | |
| GenBank mRNA | AB093256.1 | - | 8 - 9 | 1603 | 60 | |
| AK036540.1 | - | 10 - 11 | 2040 | 60 | ||
| AK145708.1 | - | 10 - 11 | 2098 | 60 | ||
| AK150045.1 | - | 10 - 11 | 2016 | 60 | ||
| AK154146.1 | - | 10 - 11 | 2045 | 60 | ||
| AK154925.1 | - | 10 - 11 | 2057 | 60 | ||
| AK157310.1 | - | 10 - 11 | 2066 | 60 | ||
| AK198311.1 | - | 1 - 2 | 267 | 60 | ||
| BC060069.1 | - | 5 - 6 | 1168 | 60 | ||
| BC094446.1 | - | 1 - 2 | 636 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cluh
Genomic Map