Assay Details
| Gene Symbol: Cluh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081158.2 | NP_001074627.1 | 3 - 4 | 576 | 114 | |
| GenBank mRNA | AB093256.1 | - | 1 - 2 | 123 | 114 | |
| AK036540.1 | - | 3 - 4 | 560 | 114 | ||
| AK145708.1 | - | 3 - 4 | 617 | 114 | ||
| AK150045.1 | - | 3 - 4 | 535 | 114 | ||
| AK154146.1 | - | 3 - 4 | 565 | 114 | ||
| AK154925.1 | - | 3 - 4 | 577 | 114 | ||
| AK157310.1 | - | 3 - 4 | 586 | 114 | ||
| BC072573.1 | - | 3 - 4 | 531 | 5705116 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cluh
Genomic Map