Assay Details
| Gene Symbol: Agt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007428.3 | NP_031454.3 | 2 - 3 | 887 | 65 | |
| GenBank mRNA | AK018763.1 | - | 2 - 3 | 885 | 65 | |
| AK139179.1 | - | 2 - 3 | 886 | 65 | ||
| BC019496.1 | - | 1 - 2 | 863 | 4211501 | 65 | |
| BC028877.1 | - | 1 - 2 | 853 | 4194190 | 65 | |
| BC054387.1 | - | 872 | 6477694 | 65 | ||
More Information
Set Membership: |
|

Agt
Genomic Map