Assay Details
| Gene Symbol: Galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139272.2 | NP_644678.2 | 9 - 10 | 1033 | 94 | |
| GenBank mRNA | AF348968.1 | - | 9 - 10 | 901 | 94 | |
| AK046567.1 | - | 9 - 10 | 991 | 94 | ||
| AK145122.1 | - | 9 - 10 | 911 | 94 | ||
| AK149609.1 | - | 9 - 10 | 961 | 94 | ||
| AK151536.1 | - | 9 - 10 | 824 | 94 | ||
| BC007172.1 | - | 8 - 9 | 768 | 94 | ||
| BC053063.1 | - | 8 - 9 | 876 | 5702572 | 94 | |
| BC059818.1 | - | 9 - 10 | 978 | 5697789 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt2
Genomic Map