Assay Details
| Gene Symbol: Nqo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008706.5 | NP_032732.3 | 3 - 4 | 438 | 76 | |
| GenBank mRNA | AK075568.1 | - | 3 - 4 | 439 | 76 | |
| BC004579.1 | - | 3 - 4 | 416 | 3586888 | 76 | |
| CT010284.1 | - | 2 - 3 | 305 | 76 | ||
| S75951.1 | - | 3 - 4 | 512 | 76 | ||
| U12961.1 | - | 3 - 4 | 413 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nqo1
Genomic Map