Assay Details
| Gene Symbol: Ces1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021456.4 | NP_067431.2 | 3 - 4 | 457 | 66 | |
| GenBank mRNA | AI316467.1 | - | 4 - 5 | 338 | 66 | |
| AK136446.1 | - | 3 - 4 | 454 | 66 | ||
| AK136588.1 | - | 3 - 4 | 454 | 66 | ||
| AK149446.1 | - | 3 - 4 | 454 | 66 | ||
| AK162328.1 | - | 3 - 4 | 432 | 66 | ||
| BC021150.1 | - | 3 - 4 | 476 | 4163576 | 66 | |
| BC026897.1 | - | 3 - 4 | 437 | 4195247 | 66 | |
| Y12887.1 | - | 3 - 4 | 443 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ces1g
Genomic Map