Assay Details
| Gene Symbol: Ces1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007954.4 | NP_031980.2 | 6 - 7 | 811 | 97 | |
| GenBank mRNA | AF034435.1 | - | 793 | 97 | ||
| AI527022.1 | - | 2 - 3 | 174 | 97 | ||
| AK050444.1 | - | 6 - 7 | 812 | 97 | ||
| BC028907.1 | - | 6 - 7 | 789 | 4196339 | 97 | |
| M57960.1 | - | 6 - 7 | 804 | 97 | ||
More Information
Set Membership: |
|

Ces1c
Genomic Map