Assay Details
| Gene Symbol: Slc6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY188506.1 | - | 4 - 5 | 803 | 57 | |
| AY525777.1 | - | 4 - 5 | 1100 | 57 | ||
| L19564.1 | - | 4 - 5 | 555 | 57 | ||
| U76306.1 | - | 4 - 5 | 785 | 57 | ||
More Information
Set Membership: |
|

Slc6a2
Genomic Map