Assay Details
| Gene Symbol: Slc6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009209.3 | NP_033235.3 | 6 - 7 | 1220 | 80 | |
| GenBank mRNA | AJ439987.1 | - | 6 - 7 | 1037 | 80 | |
| AY188506.1 | - | 6 - 7 | 1037 | 80 | ||
| AY525777.1 | - | 6 - 7 | 1334 | 80 | ||
| AY525778.1 | - | 6 - 7 | 1334 | 80 | ||
| BC066216.1 | - | 7 - 8 | 1154 | 30431573 | 80 | |
| U76306.1 | - | 6 - 7 | 1019 | 80 | ||
More Information
Set Membership: |
|

Slc6a2
Genomic Map