Assay Details
| Gene Symbol: Mmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008610.2 | NP_032636.1 | 4 - 5 | 958 | 57 | |
| GenBank mRNA | AK148184.1 | - | 4 - 5 | 957 | 57 | |
| AK208377.1 | - | 1 - 2 | 72 | 57 | ||
| BC070430.1 | - | 4 - 5 | 958 | 6813184 | 57 | |
| M84324.1 | - | 4 - 5 | 704 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp2
Genomic Map