Assay Details
| Gene Symbol: Trub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028115.3 | NP_082391.1 | 3 - 4 | 440 | 82 | |
| NM_029839.3 | NP_084115.2 | 3 - 4 | 440 | 82 | ||
| GenBank mRNA | AK011362.1 | - | 3 - 4 | 435 | 82 | |
| AK018525.1 | - | 3 - 4 | 413 | 82 | ||
| AK031674.1 | - | 3 - 4 | 418 | 82 | ||
| AK164095.1 | - | 3 - 4 | 411 | 82 | ||
| AK169221.1 | - | 3 - 4 | 406 | 82 | ||
| BC039287.1 | - | 3 - 4 | 385 | 82 | ||
| BC054088.1 | - | 3 - 4 | 254 | 82 | ||
| BC089592.1 | - | 3 - 4 | 407 | 6757213 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trub1
Genomic Map