Assay Details
| Gene Symbol: Trub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028115.3 | NP_082391.1 | 4 - 5 | 526 | 79 | |
| NM_029839.3 | NP_084115.2 | 4 - 5 | 526 | 79 | ||
| GenBank mRNA | AK011362.1 | - | 4 - 5 | 521 | 79 | |
| AK018525.1 | - | 4 - 5 | 499 | 79 | ||
| AK031674.1 | - | 4 - 5 | 504 | 79 | ||
| AK164095.1 | - | 4 - 5 | 497 | 79 | ||
| AK169221.1 | - | 4 - 5 | 492 | 79 | ||
| BC039287.1 | - | 4 - 5 | 471 | 79 | ||
| BC054088.1 | - | 4 - 5 | 340 | 79 | ||
| BC089592.1 | - | 4 - 5 | 493 | 6757213 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trub1
Genomic Map