Assay Details
| Gene Symbol: Pnliprp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018874.2 | NP_061362.1 | 10 - 11 | 1218 | 78 | |
| GenBank mRNA | AF061274.1 | - | 10 - 11 | 1219 | 78 | |
| AK028105.1 | - | 10 - 11 | 1131 | 78 | ||
| BC068266.1 | - | 6 - 7 | 78 | |||
| BC090985.1 | - | 10 - 11 | 1130 | 30315884 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnliprp1
Genomic Map