Assay Details
| Gene Symbol: Eno4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178689.4 | NP_848804.2 | 2 - 3 | 326 | 84 | |
| GenBank mRNA | AK032395.1 | - | 2 - 3 | 329 | 84 | |
| BB614863.1 | - | 2 - 3 | 349 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eno4
Genomic Map