Assay Details
| Gene Symbol: Sh3gl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019535.2 | NP_062408.2 | 6 - 7 | 735 | 81 | |
| GenBank mRNA | AF326561.1 | - | 6 - 7 | 614 | 81 | |
| AF326562.1 | - | 6 - 7 | 614 | 81 | ||
| AK035210.1 | - | 1066 | 81 | |||
| AK043424.1 | - | 723 | 81 | |||
| AK044370.1 | - | 6 - 7 | 750 | 81 | ||
| AK134970.1 | - | 4 - 5 | 446 | 81 | ||
| AK139337.1 | - | 6 - 7 | 700 | 81 | ||
| BC018385.1 | - | 6 - 7 | 735 | 4503038 | 81 | |
| U58886.1 | - | 6 - 7 | 698 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3gl2
Genomic Map