Assay Details
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290645.1 | NP_001277574.1 | 5 - 6 | 802 | 140 | |
NM_146148.2 | NP_666260.1 | 6 - 7 | 899 | 140 | ||
GenBank mRNA | AB077295.1 | - | 6 - 7 | 913 | 140 | |
AB077296.1 | - | 6 - 7 | 862 | 140 | ||
AB077297.1 | - | 5 - 6 | 768 | 140 | ||
AB077298.1 | - | 6 - 7 | 862 | 140 | ||
AK090245.1 | - | 6 - 7 | 898 | 140 | ||
BC027748.1 | - | 6 - 7 | 895 | 5052412 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
