| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400007454.mm.1 |
NM_001162980.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007454.mm.1 |
NM_001162980.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007454.mm.1 |
NM_001285952.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007454.mm.1 |
NM_001285953.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007454.mm.1 |
NR_104376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007454.mm.1 |
TC0400001090.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400007456.mm.1 |
NM_001162980.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007456.mm.1 |
NM_001162980.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007456.mm.1 |
NM_001285952.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007456.mm.1 |
NM_001285953.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007456.mm.1 |
NR_104376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007456.mm.1 |
TC0400001090.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400007455.mm.1 |
NM_001162980.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007455.mm.1 |
NM_001162980.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007455.mm.1 |
NM_001285952.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007455.mm.1 |
NM_001285953.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007455.mm.1 |
NR_104376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007455.mm.1 |
TC0400001090.mm.1 |
MTA TC |