Assay Details
| Gene Symbol: Smarcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018817.2 | NP_061287.2 | 15 - 16 | 2654 | 110 | |
| GenBank mRNA | AF088884.1 | - | 15 - 16 | 2634 | 110 | |
| AK043129.1 | - | 4 - 5 | 469 | 110 | ||
| AK083488.1 | - | 15 - 16 | 2632 | 110 | ||
| AK154020.1 | - | 15 - 16 | 2637 | 110 | ||
| BC029078.1 | - | 15 - 16 | 2589 | 3992325 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smarcal1
Genomic Map