Assay Details
| Gene Symbol: Rpl37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009084.4 | NP_033110.1 | 1 - 2 | 64 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_009084.4 transcripts.
Set Membership: |
|

Rpl37a
Genomic Map