Assay Details
| Gene Symbol: 1700027A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038001.1 | 1 - 2 | 57 | 89 | ||
| GenBank mRNA | AK006406.1 | - | 1 - 2 | 58 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transition protein 1 | Tnp1 |
Stp-1, TP1, Tp-1 |
-137.0 | Anti-sense | Chr. 1 - 73015074 - 73015899 on NCBI Build GRCm38 |
| Downstream | pregnancy induced noncoding RNA | Pinc | 373.8kb | Anti-sense | Chr. 1 - 73391385 - 73407569 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700027A15Rik
Genomic Map