Assay Details
| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177980.1 | NP_001171451.1 | 6 - 7 | 1455 | 118 | |
| NM_001177981.1 | NP_001171452.1 | 6 - 7 | 995 | 118 | ||
| NM_001177982.1 | NP_001171453.1 | 6 - 7 | 731 | 118 | ||
| NM_019840.2 | NP_062814.2 | 11 - 12 | 1620 | 118 | ||
| GenBank mRNA | AF208023.2 | - | 11 - 12 | 1555 | 118 | |
| AF326555.1 | - | 11 - 12 | 1360 | 118 | ||
| AF326556.1 | - | 11 - 12 | 1360 | 118 | ||
| AJ297397.1 | - | 11 - 12 | 1555 | 118 | ||
| AK051102.1 | - | 6 - 7 | 996 | 118 | ||
| AK161341.1 | - | 6 - 7 | 732 | 118 | ||
| BC007155.1 | - | 6 - 7 | 802 | 118 | ||
| BC023751.1 | - | 6 - 7 | 1431 | 118 | ||
| BC071259.1 | - | 6 - 7 | 971 | 5368075 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde4b
Genomic Map