Assay Details
| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133810.3 | NP_598571.2 | 5 - 6 | 791 | 67 | |
| GenBank mRNA | AK029551.1 | - | 5 - 6 | 734 | 67 | |
| AK035693.1 | - | 5 - 6 | 759 | 67 | ||
| AK041139.1 | - | 1 - 2 | 126 | 67 | ||
| AK076932.1 | - | 5 - 6 | 758 | 67 | ||
| AK083332.1 | - | 5 - 6 | 760 | 67 | ||
| AK153108.1 | - | 5 - 6 | 735 | 67 | ||
| AK165291.1 | - | 5 - 6 | 731 | 67 | ||
| AY092028.1 | - | 5 - 6 | 744 | 67 | ||
| BC006579.1 | - | 5 - 6 | 746 | 3586767 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk17b
Genomic Map