Assay Details
| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133810.3 | NP_598571.2 | 6 - 7 | 837 | 131 | |
| GenBank mRNA | AK029551.1 | - | 6 - 7 | 780 | 131 | |
| AK035693.1 | - | 6 - 7 | 805 | 131 | ||
| AK041139.1 | - | 2 - 3 | 172 | 131 | ||
| AK076932.1 | - | 6 - 7 | 804 | 131 | ||
| AK083332.1 | - | 6 - 7 | 806 | 131 | ||
| AK153108.1 | - | 6 - 7 | 781 | 131 | ||
| AK165291.1 | - | 6 - 7 | 777 | 131 | ||
| AY092028.1 | - | 6 - 7 | 790 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk17b
Genomic Map