Assay Details
| Gene Symbol: Gtf3c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033194.2 | NP_001028366.1 | 5 - 6 | 776 | 108 | |
| GenBank mRNA | AK145040.1 | - | 777 | 108 | ||
| AK160997.1 | - | 5 - 6 | 764 | 108 | ||
| AK165830.1 | - | 5 - 6 | 777 | 108 | ||
| BC141157.1 | - | 5 - 6 | 766 | 9055769 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c3
Genomic Map