Assay Details
| Gene Symbol: Pgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163314.2 | NP_001156786.1 | 12 - 13 | 1377 | 81 | |
| GenBank mRNA | AK138171.1 | - | 12 - 13 | 1317 | 81 | |
More Information
Set Membership: |
|

Pgap1
Genomic Map