Assay Details
| Gene Symbol: Pgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163314.2 | NP_001156786.1 | 15 - 16 | 1596 | 135 | |
| GenBank mRNA | AK138171.1 | - | 15 - 16 | 1536 | 135 | |
| CK392830.1 | - | 2 - 3 | 130 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pgap1
Genomic Map