| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100019013.mm.1 |
NM_001163314.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019013.mm.1 |
NM_001163314.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019013.mm.1 |
TC0100002349.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100019015.mm.1 |
NM_001163314.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019015.mm.1 |
NM_001163314.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019015.mm.1 |
TC0100002349.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100019014.mm.1 |
NM_001163314.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019014.mm.1 |
NM_001163314.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019014.mm.1 |
TC0100002349.mm.1 |
MTA TC |