| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100018995.mm.1 |
NM_001163314.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018995.mm.1 |
NM_001163314.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018995.mm.1 |
TC0100002349.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100018993.mm.1 |
NM_001163314.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018993.mm.1 |
NM_001163314.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018993.mm.1 |
TC0100002349.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100018994.mm.1 |
NM_001163314.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018994.mm.1 |
NM_001163314.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018994.mm.1 |
TC0100002349.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100018996.mm.1 |
NM_001163314.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018996.mm.1 |
NM_001163314.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018996.mm.1 |
TC0100002349.mm.1 |
MTA TC |