Assay Details
| Gene Symbol: Ctla4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009843.4 | NP_033973.2 | 3 - 4 | 708 | 66 | |
| GenBank mRNA | AK138891.1 | - | 3 - 4 | 642 | 66 | |
| BC042741.1 | - | 3 - 4 | 708 | 4936052 | 66 | |
| BC052683.1 | - | 3 - 4 | 628 | 30072821 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctla4
Genomic Map