Assay Details
| Gene Symbol: Tdrd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028034.2 | NP_082310.1 | 1 - 2 | 226 | 159 | |
| GenBank mRNA | AB211061.1 | - | 1 - 2 | 165 | 159 | |
| AK010386.1 | - | 1 - 2 | 165 | 159 | ||
| AK135918.1 | - | 1 - 2 | 146 | 159 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB211061.1, AK010386.1, NM_028034.2 transcripts.
Set Membership: |
|

Tdrd12
Genomic Map