Assay Details
| Gene Symbol: Tdrd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028034.2 | NP_082310.1 | 9 - 10 | 1158 | 90 | |
| GenBank mRNA | AB211061.1 | - | 9 - 10 | 1097 | 90 | |
| AK010386.1 | - | 9 - 10 | 1097 | 90 | ||
| AK010714.1 | - | 8 - 9 | 947 | 90 | ||
| AK135918.1 | - | 9 - 10 | 1079 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tdrd12
Genomic Map