Assay Details
| Gene Symbol: Rhpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027897.4 | NP_082173.3 | 1 - 2 | 172 | 102 | |
| GenBank mRNA | AK004849.1 | - | 1 - 2 | 95 | 102 | |
| AK050214.1 | - | 1 - 2 | 86 | 102 | ||
| BC100314.1 | - | 1 - 2 | 79 | 30848276 | 102 | |
| CJ080543.1 | - | 1 - 2 | 172 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004849.1, AK050214.1, BC100314.1, NM_027897.4 transcripts.
Set Membership: |
|

Rhpn2
Genomic Map