Assay Details
| Gene Symbol: Fhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289533.1 | NP_001276462.1 | 3 - 4 | 461 | 64 | |
| NM_010212.4 | NP_034342.1 | 3 - 4 | 610 | 64 | ||
| GenBank mRNA | AF153340.1 | - | 2 - 3 | 382 | 64 | |
| AK052936.1 | - | 3 - 4 | 462 | 64 | ||
| AK133708.1 | - | 3 - 4 | 582 | 64 | ||
| AK147024.1 | - | 3 - 4 | 611 | 64 | ||
| BC021468.1 | - | 3 - 4 | 519 | 5066565 | 64 | |
| U77040.2 | - | 3 - 4 | 500 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fhl2
Genomic Map