Assay Details
| Gene Symbol: Mrps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023514.4 | NP_076003.3 | 3 - 4 | 436 | 132 | |
| GenBank mRNA | AK009082.1 | - | 3 - 4 | 386 | 132 | |
| AK049465.1 | - | 3 - 4 | 394 | 132 | ||
| AK144656.1 | - | 3 - 4 | 394 | 132 | ||
| BC051936.1 | - | 2 - 3 | 129 | 132 | ||
| BC083159.1 | - | 2 - 3 | 197 | 132 | ||
| BC116437.2 | - | 3 - 4 | 386 | 40104493 | 132 | |
More Information
Set Membership: |
|

Mrps9
Genomic Map