Assay Details
| Gene Symbol: Mrps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023514.4 | NP_076003.3 | 5 - 6 | 546 | 87 | |
| GenBank mRNA | AK009082.1 | - | 5 - 6 | 496 | 87 | |
| AK049465.1 | - | 5 - 6 | 504 | 87 | ||
| AK150985.1 | - | 5 - 6 | 503 | 87 | ||
| BC051936.1 | - | 4 - 5 | 239 | 87 | ||
| BC069980.1 | - | 3 - 4 | 140 | 87 | ||
| BC083159.1 | - | 4 - 5 | 307 | 87 | ||
| BC116437.2 | - | 5 - 6 | 496 | 40104493 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps9
Genomic Map