Assay Details
| Gene Symbol: Mrps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023514.4 | NP_076003.3 | 8 - 9 | 866 | 52 | |
| GenBank mRNA | AJ250193.1 | - | 4 - 5 | 335 | 52 | |
| AK009082.1 | - | 8 - 9 | 816 | 52 | ||
| AK144656.1 | - | 8 - 9 | 824 | 52 | ||
| AK150985.1 | - | 8 - 9 | 823 | 52 | ||
| BC022587.1 | - | 3 - 4 | 304 | 52 | ||
| BC051936.1 | - | 7 - 8 | 559 | 52 | ||
| BC069980.1 | - | 6 - 7 | 460 | 52 | ||
| BC116437.2 | - | 8 - 9 | 816 | 40104493 | 52 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC022587.1 transcripts.
Set Membership: |
|

Mrps9
Genomic Map