Assay Details
| Gene Symbol: Tgfbrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013025.2 | NP_001013043.1 | 11 - 12 | 2542 | 74 | |
| GenBank mRNA | AK141742.1 | - | 10 - 11 | 2098 | 74 | |
| BC030488.1 | - | 1 - 2 | 164 | 74 | ||
| BC072608.1 | - | 11 - 12 | 2542 | 74 | ||
| BC138866.1 | - | 11 - 12 | 2456 | 8861888 | 74 | |
| BC138867.1 | - | 11 - 12 | 2456 | 8861889 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgfbrap1
Genomic Map