Assay Details
| Gene Symbol: Tgfbrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013025.2 | NP_001013043.1 | 8 - 9 | 2086 | 114 | |
| GenBank mRNA | AK082918.1 | - | 2 - 3 | 1112 | 114 | |
| AK141742.1 | - | 7 - 8 | 1642 | 114 | ||
| BC072608.1 | - | 8 - 9 | 2086 | 114 | ||
| BC138866.1 | - | 8 - 9 | 2000 | 8861888 | 114 | |
| BC138867.1 | - | 8 - 9 | 2000 | 8861889 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgfbrap1
Genomic Map