Assay Details
| Gene Symbol: AI597479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133818.1 | NP_598579.1 | 2 - 3 | 322 | 142 | |
| GenBank mRNA | AK081372.1 | - | 2 - 3 | 313 | 142 | |
| BC006931.1 | - | 2 - 3 | 322 | 3598975 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

AI597479
Genomic Map