Assay Details
| Gene Symbol: Wdr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146254.4 | NP_666366.4 | 11 - 12 | 1751 | 68 | |
| GenBank mRNA | AK030997.1 | - | 4 - 5 | 3436 | 68 | |
| BC028975.1 | - | 11 - 12 | 1676 | 5363117 | 68 | |
| BC034130.1 | - | 3 - 4 | 731 | 4920514 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr78
Genomic Map