Assay Details
| Gene Symbol: Wdr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146254.4 | NP_666366.4 | 12 - 13 | 1916 | 93 | |
| GenBank mRNA | AK030997.1 | - | 5 - 6 | 3601 | 93 | |
| BC034130.1 | - | 4 - 5 | 896 | 4920514 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr78
Genomic Map