Assay Details
| Gene Symbol: Wdr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146254.4 | NP_666366.4 | 13 - 14 | 2040 | 107 | |
| GenBank mRNA | AK030997.1 | - | 6 - 7 | 3725 | 107 | |
| BC034130.1 | - | 5 - 6 | 1020 | 4920514 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr78
Genomic Map