Assay Details
| Gene Symbol: Wdr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146254.4 | NP_666366.4 | 5 - 6 | 817 | 154 | |
| GenBank mRNA | BC028975.1 | - | 5 - 6 | 742 | 5363117 | 154 |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr78
Genomic Map