Assay Details
| Gene Symbol: Taf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100449.1 | NP_001093919.1 | 10 - 11 | 2328 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taf4b| Gene Symbol: Taf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100449.1 | NP_001093919.1 | 10 - 11 | 2328 | 93 | |
Set Membership: |
|