Assay Details
| Gene Symbol: Taf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100449.1 | NP_001093919.1 | 12 - 13 | 2591 | 91 | |
| GenBank mRNA | AK029943.1 | - | 1 - 2 | 136 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf4b
Genomic Map