Assay Details
| Gene Symbol: Klf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033563.2 | NP_291041.2 | 2 - 3 | 1125 | 95 | |
| GenBank mRNA | AB221646.1 | - | 2 - 3 | 737 | 95 | |
| AF327568.1 | - | 2 - 3 | 1016 | 95 | ||
| AF338369.1 | - | 2 - 3 | 1160 | 95 | ||
| AK036514.1 | - | 1 - 2 | 123 | 95 | ||
| AK086122.1 | - | 1 - 2 | 80 | 95 | ||
| AK149362.1 | - | 3 - 4 | 916 | 95 | ||
| BC004700.1 | - | 3 - 4 | 890 | 3499191 | 95 | |
| BC019604.1 | - | 2 - 3 | 1125 | 3601534 | 95 | |
| BC040794.1 | - | 3 - 4 | 890 | 3499191 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Klf7
Genomic Map