Assay Details
| Gene Symbol: Tdrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002238.2 | NP_001002238.1 | 18 - 19 | 2551 | 99 | |
| NM_001002240.2 | NP_001002240.1 | 18 - 19 | 2524 | 99 | ||
| NM_001002241.2 | NP_001002241.1 | 17 - 18 | 2824 | 99 | ||
| NM_031387.3 | NP_113564.2 | 18 - 19 | 2684 | 99 | ||
| GenBank mRNA | AB067571.1 | - | 17 - 18 | 2505 | 99 | |
| AB183526.1 | - | 18 - 19 | 2551 | 99 | ||
| AB183527.1 | - | 18 - 19 | 2524 | 99 | ||
| AB183528.1 | - | 17 - 18 | 2824 | 99 | ||
| AB183529.1 | - | 17 - 18 | 2388 | 99 | ||
| AF285591.1 | - | 14 - 15 | 1878 | 99 | ||
| AK029806.1 | - | 18 - 19 | 2685 | 99 | ||
| BC129954.1 | - | 13 - 14 | 1810 | 40081321 | 99 | |
| BC129955.1 | - | 13 - 14 | 1810 | 40081323 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tdrd1
Genomic Map